missing translation for 'onlineSavingsMsg'
Learn More

RANBP9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 51038-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16816315

  • 319.00€ / 150 microlitros

Envío estimado: 02-11-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a protein that binds RAN, a small GTP binding protein belonging to the RAS superfamily that is essential for the translocation of RNA and proteins through the nuclear pore complex. The protein encoded by this gene has also been shown to interact with several other proteins, including met proto-oncogene, homeodomain interacting protein kinase 2, androgen receptor, and cyclin-dependent kinase 11.
Especificaciones

Especificaciones

RANBP9
0.19 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P69566, Q96S59
BPM L, BPM90, RAN binding protein 9, Ran binding protein M, RanBP7, RANBP9, RANBPM
Rabbit
IgG
150 μL
-20°C
Primary
10048, 56705
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RANBP9
Liquid
Ranbp9
RANBP9 Fusion Protein Ag0428
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Videos
SDS
Documentos
Certificados