missing translation for 'onlineSavingsMsg'
Learn More

Rad51 Antibody (RAD51/2701), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP2-79895IR

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215707

  • 578.82€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

Rad51 Monoclonal specifically detects Rad51 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

Rad51
Monoclonal
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
RAD51
Recombinant fragment of human Rad51 protein (around aa 1-134) (exact sequence is proprietary) (Uniprot: Q06609)
0.1 mL
Cancer, DNA Repair, Homologous Recombination, modENCODE Antibodies, Phospho Specific
Human
IgG2b κ
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
RAD51/2701
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
BRCC5, HRAD51, HsRAD51, HsT16930, RAD51 (S. cerevisiae) homolog (E coli RecA homolog), RAD51 homolog (RecA homolog, E. coli) (S. cerevisiae), RAD51 homolog A, RAD51ABRCA1/BRCA2-containing complex, subunit 5, RAD51L3, RecA, E. coli, homolog of, RECADNA repair protein RAD51 homolog 1, RecA-like protein, recombination protein A
Mouse
Protein A or G purified
RUO
Primary
Store at 4°C in the dark.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales