missing translation for 'onlineSavingsMsg'
Learn More

RAD23A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 51033-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16816305

  • 319.00€ / 150 microlitros

Envío estimado: 02-11-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This antibody is a rabbit polyclonal antibody raised against a fusion protein of full length human RAD23A.

The protein encoded by this gene is one of two human homologs of Saccharomyces cerevisiae Rad23, a protein involved in nucleotide excision repair (NER). This protein was shown to interact with, and elevate the nucleotide excision activity of 3-methyladenine-DNA glycosylase (MPG), which suggested a role in DNA damage recognition in base excision repair. This protein contains an N-terminal ubiquitin-like domain, which was reported to interact with 26S proteasome, as well as with ubiquitin protein ligase E6AP, and thus suggests that this protein may be involved in the ubiquitin mediated proteolytic pathway in cells.
Especificaciones

Especificaciones

RAD23A
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P54725, P54726
HHR23A, HR23A, RAD23A
Rabbit
IgG
150 μL
-20°C
Primary
19358, 361381, 5886
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RAD23A
Liquid
Rad23a
RAD23A Fusion Protein Ag0716
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documentos
Certificados