missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Eco AluRack for Upright Freezer, for Microtiterplates, 25 mm Shelf Height
Eco AluRack for upright freezers is designed to hold microtiterplates.
Marca: Tenak™ TE15580
Detalles adicionales : Peso : 1.60000kg
Descripción
- For use in upright freezers
- Racks can either be used as one plate per shelf or the plates can be stacked
- Constructed of aluminum
Especificaciones
6 x 7 | |
Upright Freezers | |
Aluminum |
630 x 135 x 159 mm | |
Microtiterplates (42) | |
25 mm |