Learn More
Tenak™ ECO AluRack for Upright Freezers, 75 mm Box Height
ECO AluRack fits into any upright freezer and can be used with all standard cryoboxes.
Marca: Tenak™ TE11274
Detalles adicionales : Peso : 1.30000kg
Description
- Made of seawater resistant aluminum
- Fits into any freezer and can be used with all standard cryoboxes
- Racks can be used with all freezer brands
- Without white cardboard boxes
- Holds is expressed by Height x Dept
Specifications
3 x 4 | |
564 x 139 x 240 mm | |
75 mm (Box) | |
Aluminum |
Seawater resistant | |
Upright Freezers | |
Boxes (12) |