Learn More
Tenak™ ECO AluRack for Upright Freezers, 100 mm Box Height, with Boxes
ECO AluRack fits into any upright freezer and can be used with all standard cryoboxes.
Marca: Tenak™ TE11302B
Detalles adicionales : Peso : 2.10000kg
Beschreibung
- Made of seawater resistant aluminum
- Fits into any freezer and can be used with all standard cryoboxes
- Racks can be used with all freezer brands
- Includes standard white cardboard boxes with 9 x 9 cell dividers
- Holds is expressed by Height x Dept
Spezifikation
2 x 4 | |
564 x 139 x 210 mm | |
100 mm (Box) | |
White cardboard boxes, 9 x 9 cell dividers |
Seawater resistant | |
Upright Freezers | |
Boxes (8) | |
Aluminum |