missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RABL3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11916-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Especificaciones
RABL3 | |
Polyclonal | |
Unconjugated | |
RABL3 | |
Rab like protein 3, RABL3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
285282, 360720, 67657 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5HYI8, Q9D4V7 | |
RABL3 | |
RABL3 Fusion Protein Ag2531 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |