missing translation for 'onlineSavingsMsg'
Learn More

RABL2B Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 11588-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16818773

  • 109.00€ / 20 microlitros

Envío estimado: 02-11-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The RABL2B protein is a member of the RAB gene family which belongs to the RAS GTPase superfamily. RABL2B is located within a subtelomeric region of 22q13.3. Multiple alternatively spliced transcript variants encoding several different isoforms have been found for this gene.
Especificaciones

Especificaciones

RABL2B
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9UNT1
Rab like protein 2B, RABL2B
Rabbit
IgG
20 μL
-20°C
Primary
11158
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RABL2B
Liquid
RABL2B
RABL2B Fusion Protein Ag2189
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documentos
Certificados