missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Golden Syrian Hamster IgG (H+L) Secondary Antibody, Rhodamine, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NB120-6780

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214420

  • 219.22€ / 1.50 miligramo
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Golden Syrian Hamster samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
Especificaciones

Especificaciones

IgG (H+L)
Polyclonal
Rhodamine
Rabbit
Multi-step
RUO
Secondary
Reconstitute with 1.0 ml deionized water (or equivalent).
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Flow Cytometry, Immunocytochemistry, Immunofluorescence
LYOPH
Flow Cytometry 1:1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000
Golden Syrian Hamster IgG whole molecule
1.5 mg
Adaptive Immunity, Immunology
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum, Hamster IgG and Hamster Serum.
Golden Syrian Hamster
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only