missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Chicken IgG F(ab')2 Secondary Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NBP1-73508

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214532

  • 333.68€ / 1.50 miligramo
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

IgG F(ab')2 Polyclonal antibody specifically detects IgG F(ab')2 in Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Especificaciones

Especificaciones

IgG F(ab')2
Polyclonal
Biotin
immunoglobulin heavy constant gamma 1 (G1m marker)
Chicken IgG F(ab')2 fragment
1.5 mg
Secondary
Reconstitute with 1.0 ml deionized water (or equivalent).
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
LYOPH
Western Blot 1:2000 - 1:10000, ELISA 1:20000 - 1:100000, Immunohistochemistry 1:1000 - 1:5000
Rabbit
Affinity Purified
RUO
No reaction was observed against Chicken IgG F(c).
Chicken
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only