missing translation for 'onlineSavingsMsg'
Learn More

RAB6B Antibody (6D12G9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP2-61824

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213244

  • 532.38€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

RAB6B Monoclonal specifically detects RAB6B in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

RAB6B
Monoclonal
1 mg/mL
Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000
RAB6B, member RAS oncogene family, ras-related protein Rab-6B, small GTPase RAB6B, small GTP-binding protein
Mouse
23.5 kDa
0.1 mL
Primary
Human, Rat
Purified
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
6D12G9
Unconjugated
PBS with 0.05% Sodium Azide
Rab6b
Purified recombinant fragment of human Rab6b (AA: 95-208) expressed in E. Coli.
Protein G purified
Signal Transduction
51560
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales