missing translation for 'onlineSavingsMsg'
Learn More

Rab5a Antibody (3A4), FITC, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NBP1-04340F

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213981

  • 521.58€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

Rab5a Monoclonal specifically detects Rab5a in Human, Mouse samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Especificaciones

Especificaciones

Rab5a
Monoclonal
FITC
PBS with 0.05% Sodium Azide
RAB5A
Recombinant human Rab5a (1-215aa) purified from E. coli
0.1 mL
Endosome Markers, Signal Transduction
5868
Human, Mouse
Purified
ELISA
3A4
ELISA
RAB5A, member RAS oncogene family, RAB5RAS-associated protein RAB5A, ras-related protein Rab-5A
Mouse
Protein G purified
RUO
Primary
Anti-human Rab5a mAb, clone 3A4, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human Rab5a protein.
Store at 4C in the dark.
IgG2b κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales