Learn More
R&D Systems™ Quantikine Immunoassay Control Set 788 Human Galectin-3BP
Product Code. 18743399
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
Product Code. Quantity Unit Size
18743399 1 Each Each
1 options
This item is not returnable. View return policy
Product Code. 18743399 Supplier R&D Systems™ Supplier No. QC95
  • ONLINE EXCLUSIVE PRICE
    221.93€
    198.45€ / paquete de 1
    Ahorro 23.48€
    10% Descuento
 Restricted Item
Fecha estimada de envoi: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For use with catalog number DGBP30
Quantikine Immunoassay Control Set 788 for human Galectin-3BP (catalog number QC95) is designed as a quantitative control for cytokine concentration determination in biological fluids.
  • Product Type: Immunoassay control set
  • Contents: Lyophilized controls containing recombinant human cytokines at low, medium, and high concentrations, prepared in diluted porcine serum with preservatives
  • Application: For research use only
  • Species Reactivity: Human
  • Storage: Store unopened product at 2°C to 8°C; stable for at least 6 months from date of receipt
  • Preparation: Reconstitute vials with deionized or distilled water as indicated on the product datasheet
  • Validated Systems: Intended for use with R&D Systems Quantikine ELISA kits; values may differ when used with other systems
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes Galectin-3BP (MAC-2BP)
Sample Type Biological Fluids
Type Immunoassay Control Set
Gene ID (Entrez) 3959
Intended Use Control
Product Line R&D Systems
Quantity 1 Each
Source Human
Form Lyophilized