Learn More
(R)-Prunasin, TRC
Product Code. 30041053
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041053 2.5 mg 2.5mg
30040798 10 mg 10mg
30040543 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041053 Supplier Toronto Research Chemicals Supplier No. TRCP8390002.5MG
 Restricted Item
Fecha estimada de envoi: 17-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 99-18-3
Molecular Formula C14 H17 N O6
Molecular Weight (g/mol) 295.29
IUPAC Name (2R)-2-phenyl-2-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyacetonitrile
SMILES OC[C@H]1O[C@@H](O[C@@H](C#N)c2ccccc2)[C@H](O)[C@@H](O)[C@@H]1O
Specifications
Quantity 2.5 mg
InChI Formula InChI=1S/C14H17NO6/c15-6-9(8-4-2-1-3-5-8)20-14-13(19)12(18)11(17)10(7-16)21-14/h1-5,9-14,16-19H,7H2/t9-,10+,11+,12-,13+,14+/m0/s1
Formula Weight 295.1056
Percent Purity >95%
Chemical Name or Material Prunasin
Grade HPLC