Learn More
IKA™ R 2722 H-Stand
Product Code. 10314681 Shop All IKA Products
Change view
Click to view available options
Height (Metric):
1.01 mm
Unit Size:
Each
Product Code. Height (Metric) Unit Size
10314681 1.01 mm Each
1 options
Product Code. 10314681 Supplier IKA™ Supplier No. 0001412000
  • ONLINE EXCLUSIVE PRICE
    1130.00€
    851.00€ / paquete de 1
    Ahorro 279.00€
    24% Descuento
En stock
Add to basket
Edge
Add to basket
Particularly stable stand with H-shape base which prevents the stand from tipping backwards.
Provides optimum stability required for larger, heavier instruments and attachments, for example with rheological measurements using overhead stirrers. The stand has an adjustment screw which can be used to compensate for an uneven laboratory table surface.
TRUSTED_SUSTAINABILITY
Specifications
Height (Metric) 1.01 mm
Length (Metric) Base 420 mm
Width (English) Base 18 in.
Width (Metric) Base 460 mm
Diameter (Metric) Rod 34 mm
Holds 10 kg
Product Type H-Stand