missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Amino-3-methylbutane, ChiPros 98%, ee 97%, Thermo Scientific™

Product Code. 11359105
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359105 25 g 25g
11339105 1 g 1g
11349105 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11359105 Supplier Thermo Scientific Alfa Aesar Supplier No. L16330.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(R)-(-)-2-Amino-3-methylbutane is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water (60g/L at 20°C).

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store under dry inert gas. Incompatible with oxidizing agents. Store in cool place. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34701-33-2
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD01075731
InChI Key JOZZAIIGWFLONA-RXMQYKEDSA-N
Synonym r---2-amino-3-methylbutane, r---3-methyl-2-butylamine, r-3-methyl-2-butylamine, r-3-methylbutan-2-amine, 2r-3-methylbutan-2-amine, unii-s9ygl88qkq, s9ygl88qkq, 2r-3-methyl-butan-2-amine, 2-butanamine, 3-methyl-, r, r-3-methyl-2-butanamine
PubChem CID 6999786
IUPAC Name (2R)-3-methylbutan-2-amine
SMILES CC(C)C(C)N

Specifications

Density 0.746
Boiling Point 85°C to 87°C
Flash Point <21°C (69°F)
Refractive Index 1.406
Quantity 25 g
UN Number UN2733
Beilstein 1718798
Sensitivity Air Sensitive
Solubility Information Soluble in water (60g/L at 20°C).
Optical Rotation −5.2° (Neat)
Formula Weight 87.16
Percent Purity 98%
Assay ee 97%
Chemical Name or Material (R)-(-)-2-Amino-3-methylbutane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.