Learn More
Quinoline-5-sulfonyl Chloride, TRC
Product Code. 30417821
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity Unit Size
30417821 1 g 1g
1 options
This item is not returnable. View return policy
Product Code. 30417821 Supplier Toronto Research Chemicals Supplier No. TRCQ7009951G
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Quinoline-5-sulfonyl Chloride
CAS 102878-84-2
Molecular Formula C9H6ClNO2S
Quantity 1 g
Synonym 5-Quinolinesulfonyl Chloride
InChI Formula InChI=1S/C9H6ClNO2S/c10-14(12,13)9-5-1-4-8-7(9)3-2-6-11-8/h1-6H
SMILES ClS(=O)(=O)c1cccc2ncccc12
IUPAC Name quinoline-5-sulfonyl chloride
Molecular Weight (g/mol) 227.67
Formula Weight 226.9808