Learn More
Quinine Di-N-oxide, TRC
Product Code. 30411647
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity Unit Size
30411647 10 mg 10mg
1 options
This item is not returnable. View return policy
Product Code. 30411647 Supplier Toronto Research Chemicals Supplier No. TRCQ69400510MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Quinine Di-N-oxide
CAS 101655-92-9
Molecular Formula C20 H24 N2 O4
Quantity 10 mg
Synonym Cinchonan-9-ol','6'-methoxy-','1,1'-dioxide','(8α,9R)-, Quinine N,N'-Dioxide (7CI), Quinine Di-N-Oxide
InChI Formula InChI=1S/C20H24N2O4/c1-3-13-12-22(25)9-7-14(13)10-19(22)20(23)16-6-8-21(24)18-5-4-15(26-2)11-17(16)18/h3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3/t13-,14-,19-,20+,22?/m0/s1
SMILES COc1ccc2c(c1)c(cc[n+]2[O-])[C@@H](O)[C@@H]3C[C@@H]4CC[N+]3([O-])C[C@@H]4C=C
IUPAC Name (R)-[(2S,4S,5R)-5-ethenyl-1-oxido-1-azoniabicyclo[2.2.2]octan-2-yl]-(6-methoxy-1-oxidoquinolin-1-ium-4-yl)methanol
Molecular Weight (g/mol) 356.42
Formula Weight 356.1736