Learn More
Quickfit™ Pear-Shaped Glass Funnel with Glass Key and Stem with Cone
For use in liquid-liquid extractions to separate (partition) the components of a mixture into two immiscible solvent phases of different densities.
Marca:  Quickfit D2/62
Detalles adicionales : Peso : 0.47000kg
 Ver más versiones de este producto
Código de producto. 15557790
  • 680.98€ / paquete de 5
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
  • Complies with ISO 4800 Type 2
  • Funnel stem with cone to allow easy connection to Quickfit™ flasks etc
  • Stopcock features interchangeable glass key
Especificaciones
Especificaciones
ISO 4800
29/32
1PK
1000mL
135mm
Separating funnel
4mm; Glass
Borosilicate Glass
Videos