Learn More
Tapón de vidrio expandido hueco hexagonal Quickfit™
Idóneas para su uso con matraces aforados.
Marca: Quickfit TRF699
Detalles adicionales : Peso : 0.43000kg
Descripción
Idóneas para su uso con matraces aforados.
- Lower closed end of stopper features drip-tip to ensure adhered liquid is returned to flask
- Hexagonal head prevents stopper from rolling and provides good grip
- Flat head allows stopper to be stood on end, thus preventing contamination of ground surface
- Hollow-blown to reduce weight
Especificaciones
5/Pk. | |
Borosilicate Glass | |
Volumetric flasks | |
45/40 |