Learn More
Quickfit™ Stirrer Guides
Designed to reduce excessive whip in glass stirrer shafts which can lead to breakage.
Marca: Quickfit ST2/44C/SC
Detalles adicionales : Peso : 0.12000kg
Descripción
- PTFE bearing is fitted to the bottom of guides ST2/33C/SC and ST2/44C/SC for additional support
- Features a side-arm with detachable plastic connector for the introduction of gases or connection to a vacuum supply
Especificaciones
To reduce excessive 'whip' in Glass stirrer shafts which can lead to breakage | |
Detachable Plastic connector, PTFE bearing | |
300 mm | |
29/32 |