missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Chromatography Column with Ground Cone

Manufactured from borosilicate glass. Quickfit™ Chromatography Column with Ground Cone includes a 0 porosity chemically resistant glass sintered disc. Upper socket can be connected to a solvent reservoir. Lower cone can be connected to stopcock adapter CR3T/23 to control eluent flow.

Marca:  Quickfit CR32/30

Detalles adicionales : Peso : 0.15000kg

 Ver más versiones de este producto

Código de producto. 15547770

  • 492.54€ / paquete de 5


Consúltenos : 902 239 303 
Añadir a la cesta

Descripción

Descripción

  • With glass sintered disc - porosity 0 to support column packing
  • Upper socket can be connected to a solvent reservoir
  • Lower cone can be connected to stopcock adapter CR3T/23 to control eluent flow
  • Manufactured from borosilicate glass
Especificaciones

Especificaciones

Chromatography column
415mm
19/26 Cone, 24/29 Socket
Glass sintered disc
1PK
Pyrex™ Borosilicate Glass
18mm
Videos