Learn More
- Home
- Products
- Flasks
- Boiling Flasks
- Quickfit™ Three-Neck Round-Bottom Glass Flask
Quickfit™ Three-Neck Round-Bottom Glass Flask
Product Code. 12030673 Shop All Masterflex Products
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
3000 mL
Socket Joint:
14/23 (Centre), 14/23 (side)
19/26 (Centre), 14/23 (side)
19/26 (Centre), 19/26 (side)
24/29 (Centre), 14/23 (side)
24/29 (Centre), 19/26 (side)
29/32
29/32 (Centre), 14/23 (side)
29/32 (Centre), 19/26 (side)
29/32 (Centre), 29/32 (side)
34/35 (Centre), 19/26 (side)
Unit Size:
Each
Pack of 5
23 product options available for selection
| Product Code. | Capacity (Metric) | Socket Joint | Unit Size |
|---|---|---|---|
| 12030673 | 500 mL | 24/29 (Centre), 19/26 (side) | Each |
| 11710259 | 100 mL | 24/29 (Centre), 14/23 (side) | Each |
| 10228054 | 1000 mL | 24/29 (Centre), 19/26 (side) | Each |
| 12020643 | 100 mL | 24/29 (Centre), 14/23 (side) | Each |
| 11301945 | 100 mL | 19/26 (Centre), 14/23 (side) | Each |
| 12488966 | 100 mL | 24/29 (Centre), 19/26 (side) | Each |
| 11371955 | 250 mL | 19/26 (Centre), 19/26 (side) | Each |
| 12408996 | 250 mL | 24/29 (Centre), 14/23 (side) | Each |
| 12622012 | 250 mL | 29/32 (Centre), 19/26 (side) | Each |
| 12591738 | 25 mL | 14/23 (Centre), 14/23 (side) | Each |
| 11900895 | 500 mL | 29/32 (Centre), 19/26 (side) | Each |
| 12493621 | 50 mL | 14/23 (Centre), 14/23 (side) | Each |
| 12413631 | 50 mL | 19/26 (Centre), 14/23 (side) | Each |
| 15557860 | 1000 mL | 29/32 | Pack of 5 |
| 12050653 | 250 mL | 24/29 (Centre), 19/26 (side) | Each |
| 15577860 | 1000 mL | 29/32 (Centre), 29/32 (side) | Pack of 5 |
| 15587860 | 1000 mL | 34/35 (Centre), 19/26 (side) | Pack of 5 |
| 15547870 | 250 mL | 29/32 (Centre), 14/23 (side) | Pack of 5 |
| 15577870 | 2000 mL | 24/29 (Centre), 19/26 (side) | Pack of 5 |
| 15527880 | 2000 mL | 34/35 (Centre), 19/26 (side) | Pack of 5 |
| 15597880 | 3000 mL | 34/35 (Centre), 19/26 (side) | Pack of 5 |
| 15567890 | 50 mL | 19/26 (Centre), 19/26 (side) | Pack of 5 |
| 15587890 | 500 mL | 24/29 (Centre), 14/23 (side) | Pack of 5 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
This item is not returnable.
View return policy
Product Code.
12030673
Supplier
Quickfit™
Supplier No.
FR500/3S/22A
-
69.14€
58.93€ / paquete de 1
Ahorro 10.21€
14% Descuento
En stock
Inicie sesión para ver el stock
Inicie sesión para ver el stock
Add to basket
Add to basket
This item is not returnable.
View return policy
Description
Useful for introducing gas inlet tubes and thermometers into the flask. The Quickfit Jointed Ware™ Quickfit™ Three-Neck Round-Bottom Flask has precision-ground sockets from borosilicate glass.
- With highest-quality Quickfit™ ground glass socket
- Side neck useful for introducing gas inlet tubes and thermometers etc., into the flask
- Manufactured from borosilicate glass for highest resistance to thermal shock and chemical attack
Specifications
Specifications
| Certifications/Compliance | ISO 383 |
| Capacity (Metric) | 500 mL |
| Material | Borosilicate Glass |
| Height (Metric) | 150 mm |
| No. per Case | 3 |
| Socket Joint | 24/29 (Centre), 19/26 (side) |
| Bottom Shape | Round |