Learn More
Quickfit™ Borosilicate Glass Soxhlet Extractor
Product Code. 11381885
Change view
Click to view available options
Capacity (Metric):
20 mL
40 mL
60 mL
100 mL
200 mL
Standard Taper Joint Bottom:
24/29
29/32
34/35
40/38
50/42
Height (Metric):
175 mm
200 mm
205 mm
220 mm
240 mm
250 mm
Cone Size:
19/26
24/29
34/35
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Standard Taper Joint Bottom Height (Metric) Cone Size Unit Size
11381885 200 mL 50/42 250 mm 24/29 Each
11371885 100 mL 40/38 220 mm 24/29 Each
12004793 100 mL 34/35 240 mm 34/35 Each
11331885 20 mL 24/29 175 mm 19/26 Each
11351885 60 mL 34/35 205 mm 24/29 Each
11341885 40 mL 29/32 200 mm 24/29 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11381885 Supplier Quickfit Supplier No. EX5/83
  • ONLINE EXCLUSIVE PRICE
    169.05€
    127.68€ / paquete de 1
    Ahorro 41.37€
    24% Descuento
View all promotions
Fecha estimada de envoi: 07-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Conforms to BS 2071. Quickfit™ Borosilicate Glass Soxhlet Extractor removes soluble components or impurities from solids via repeated soaking/siphoning with solvent. A wide choice of extractors is available to accommodate most sizes of sample and thimbles. Accurately formed concentric siphon tube provides reliable performance.
  • Conforms to BS 2071
  • For extracting soluble components or impurities from solids via repeated soaking/siphoning with solvent
  • Wide choice of extractors to accommodate most sizes of sample and thimbles
  • Accurately formed concentric siphon tube for reliable performance
Specifications
Product Type Soxhlet Extractor
Capacity (Metric) 200 mL
Material Pyrex Borosilicate Glass
Cone Size 24/29
For Use With (Application) Ideal for extracting soluble components or impurities from solids via repeated soaking/siphoning with solvent
Certifications/Compliance BS 2071
Standard Taper Joint Bottom 50/42
Height (Metric) 250 mm