Learn More
Horst™ Quick Clip Closure
Product Code. 10541742
Change view
Click to view available options
For Use With:
Tubes > 60 mm dia.
Unit Size:
Pack of 10
Product Code. For Use With Unit Size
10541742 Tubes > 60 mm dia. Pack of 10
1 options
This item is not returnable. View return policy
Product Code. 10541742 Supplier Horst™ Supplier No. 101203
  • ONLINE EXCLUSIVE PRICE
    129.93€
    81.43€ / paquete de 10
    Ahorro 48.50€
    37% Descuento
Fecha estimada de envoi: 07-12-2026