Learn More
Thermo Scientific™ QuEChERS Primary/Secondary Amine Bulk Sorbent
Product Code. 17379872 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. Quantity Unit Size
17379872 100 g 100g
1 options
This item is not returnable. View return policy
Product Code. 17379872 Supplier Thermo Scientific™ Supplier No. 80020429100
 Restricted Item
Fecha estimada de envoi: 22-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Bulk sorbent for use in QuEChERS columns.
Bulk sorbent for use in QuEChERS columns.
TRUSTED_SUSTAINABILITY
Specifications
Type QuEChERS Clean-up Kit
Quantity 100 g