Learn More
R&D Systems™ Quantikine Immunoassay Control Set 848 for Human Glypican 3
Product Code. 18777218
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
Product Code. Quantity Unit Size
18777218 1 Each Each
1 options
This item is not returnable. View return policy
Product Code. 18777218 Supplier R&D Systems™ Supplier No. QC203
  • ONLINE EXCLUSIVE PRICE
    221.93€
    198.45€ / paquete de 1
    Ahorro 23.48€
    10% Descuento
 Restricted Item
Fecha estimada de envoi: 20-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For use with catalog number DGLY30
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes Glypican 3
Gene ID (Entrez) 2719
Intended Use Control
Product Line R&D Systems
Quantity 1 Each
Source Human