Learn More
R&D Systems™ Quantikine Immunoassay Control Set 248 for Human TNF-alpha
Product Code. 18753529
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
1 set
Product Code. Quantity Unit Size
18753529 1 Each 1 set
1 options
This item is not returnable. View return policy
Product Code. 18753529 Supplier R&D Systems™ Supplier No. QC248
  • ONLINE EXCLUSIVE PRICE
    221.93€
    198.45€ / 1 sets
    Ahorro 23.48€
    10% Descuento
 Restricted Item
Fecha estimada de envoi: 18-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For use with catalog numbers DTA00D, STA00D, and PDTA00D
For use as quantitative controls for the determination of cytokine concentrations in biological fluids. Concentrations have been assigned using R&D Systems™’ Quantikine™ ELISA kits. Controls are prepared in a buffered protein base with preservatives. They contain recombinant human cytokines at low, medium and high concentrations. Controls are supplied lyophilized.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2-8°C. Do not use past expiration date.
Detectable Analytes TNF-alpha
Type Immunoassay Control
Gene ID (Entrez) 7124
Product Line R&D Systems
Quantity 1 Each