Learn More
Invitrogen™ QuantiGene™ Sample Processing Kit, blood samples
Product Code. 32044639
Change view
Click to view available options
Quantity:
10 Plates
2 Plates
5 x 10 Plates
Unit Size:
Each
Pack of 10
Pack of 2
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32044639 2 Plates Pack of 2
32045382 10 Plates Pack of 10
31942694 5 x 10 Plates Each
32044491 5 x 10 Plates Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 32044639 Supplier Invitrogen™ Supplier No. QS0110
 Restricted Item
Fecha estimada de envoi: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
This QuantiGene Sample Processing Kit for blood samples contains reagents and instructions for the preparation of cell lysates from whole blood samples for use in QuantiGene Singleplex and QuantiGene Plex assays for RNA or DNA targets.
This QuantiGene Sample Processing Kit for blood samples contains reagents and instructions for the preparation of cell lysates from whole blood samples for use in QuantiGene Singleplex and QuantiGene Plex assays for RNA or DNA targets.
  • Easy to follow instructions
  • Preserves the integrity of the RNA/DNA without degradation
  • Choose the method that matches your whole blood or dried blood spot sample
What you get
The kit contains our proprietary lysis mixture, proteinase K, and detailed instructions for preparing whole blood from heparin, citrate, or EDTA tubes, PAXgene blood RNA tubes, Tempus™ blood RNA tubes, or from dried blood spots prepared from standard anticoagulant venous blood or finger sticks.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage • 10 mL Lysis Mixture, store at 15°C to 30°C
• 625 μL Proteinase K (50 μg/μL), store at -20°C
Format Kit
Sample Type Blood
Product Line QuantiGene
Product Type Sample Processing Kit
Quantity 2 Plates
Shipping Condition Blue Ice