missing translation for 'onlineSavingsMsg'
Learn More

Piridina, ACS, 99,0 % mín, Thermo Scientific Chemicals

Catalog Number p-4408286
Click to view available options
Cantidad:
500 mL
1 L
4 L
4 x 1 L
110-86-1
C5H5N
79.102
MFCD00011732
JUJWROOIHBZHMG-UHFFFAOYSA-N
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
1049
CHEBI:16227
piridina
C1=CC=NC=C1
110-86-1
C5H5N
79.102
MFCD00011732
JUJWROOIHBZHMG-UHFFFAOYSA-N
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
1049
CHEBI:16227
piridina
C1=CC=NC=C1

Maximum level of impurities: Evaporation residue 0.002%, H2O 0.1%, Cl 0.001%, SO4 0.001%, NH3 0.002%, Cu 5ppm, Reducing substances P.T., Solubility in water P.T.

As polar, basic, low-reactive solvent, precursor, and reagent, pyridine finds wide range of applications in pharmaceuticals, agrochemicals and in the manufacture of dyes and rubber. Further it is used in the textile industry to improve network capacity of cotton. Pyridine finds applications in specialty chemical reagents like Cornforth reagent (pyridinium dichromate, PDC), pyridinium chlorochromate (PCC), the Collins reagent (complex of chromium(VI) oxide with pyridine) and the Sarret reagent (complex of chromium(VI) oxide with pyridine). Pyridine is a widely used ligand in coordination chemistry. Pyridine-base adducts like pyridine-borane (C5H5NBH3) and pyridine-sulfur trioxide (C5H5NSO3) are widely used owing to their superior characteristics in their reactivity and solubility characteristics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Pyridine
Punto de fusión °C to 42°C
Nombre de nota For ACS analysis
Color Incolora
Aspecto Colorless liquid
Densidad 0.978
Punto de ebullición 115°C
Cantidad 500 mL
Temperatura de inflamación 20°C (68°F)
Punto de ignición 482°C (900°F)
Olor Unpleasant
Forma física Líquido
Índice de refracción 1.51
Intervalo de porcentaje de ensayo 99.0% min.
Beilstein 103233
Información de solubilidad Miscible with water,alcohol,ether,pet ether,and oils.
Formula Weight (peso de la fórmula) 79.1
Grado ACS
Mostrar más Mostrar menos

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'