Learn More
Disco sinterizado/filtro Pyrex™
Para el soplado de vidrio en la construcción de aparatos especiales
Marca: Pyrex 3090/11M
Detalles adicionales : Peso : 0.08000kg
Descripción
- Complies with ISO 4793
- Manufactured from Pyrex borosilicate glass for resistance to heat and chemical corrosion
- Close control of pore size
- Accurate and consistent disc dimensions
- High mechanical strength to withstand full vacuum
- Convenient packaging
Especificaciones
Borosilicate Glass | |
Resistance to heat and chemical corrosion | |
90 mm |
1 μm | |
Filter Disc |