Learn More
Disco sinterizado/filtro Pyrex™
Para el soplado de vidrio en la construcción de aparatos especiales
Brand: Pyrex 3020/12M
Additional Details : Peso : 0.02000kg
Description
- Complies with ISO 4793
- Manufactured from Pyrex borosilicate glass for resistance to heat and chemical corrosion
- Close control of pore size
- Accurate and consistent disc dimensions
- High mechanical strength to withstand full vacuum
- Convenient packaging
Specifications
Borosilicate Glass | |
2 | |
20 mm |
Resistance to heat and chemical corrosion | |
Filter Disc |