Learn More
Pyrex™ Graduated Dropping Funnel with Glass Key
Product Code. 11303684
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11303684 100 mL Each
11333684 1000 mL Each
11313684 250 mL Each
11323684 500 mL Each
12433481 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11303684 Supplier Pyrex™ Supplier No. 3590/02
  • ONLINE EXCLUSIVE PRICE
    130.20€
    110.55€ / paquete de 1
    Ahorro 19.65€
    15% Descuento
Fecha estimada de envoi: 15-03-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Used to transfer fluids.
  • Complies with ISO 4800 type 4
  • Manufactured from Pyrex borosilicate glass
  • Double graduated scale showing approximate volume contained/dispensed
  • Graduations in white enamel
  • With interchangeable glass key
  • Supplied with glass stopper
Specifications
Capacity (Metric) 100 mL
Material Borosilicate Glass
Graduated Yes
Includes Glass stopper and key
Graduations 2mL
Product Type Dropping Funnel