Learn More
Quickfit™ Borosilicate Glass Büchner Vacuum Filtration Funnels with Glass Sintered Disc
Useful where conventional filter papers would be destroyed by acids and alkalis
Marca:  Quickfit VB30/06M
Detalles adicionales : Peso : 0.15000kg
 Ver más versiones de este producto
Código de producto. 15558000
  • 284.56€ / paquete de 5
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
  • Büchner vacuum filtration funnel with glass sintered disc
  • Manufactured from borosilicate glass for highest resistance to chemical attack
  • Useful where conventional filter papers would be destroyed by acids and alkalis
  • With a Quickfit™ ground glass cone to allow direct filtration into Quickfit™ Büchner flasks
Especificaciones
Especificaciones
ISO 3585, DIN 12217, ASTM E-438
30 mL
Borosilicate Glass
Quickfit™ ground Glass cone