Learn More
Trajan Scientific and Medical™ Push-Pull Syringe Valve
Product Code. 10157020
Change view
Click to view available options
For Use With (Equipment):
25 μL to 2.5 mL Removable Needle Syringes
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
10157020 25 μL to 2.5 mL Removable Needle Syringes Each
1 options
This item is not returnable. View return policy
Product Code. 10157020 Supplier Trajan Scientific and Medical™ Supplier No. 031905
Fecha estimada de envoi: 07-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
An SGE syringe pre-fitted with a valve will pressurize a sample prior to injection.
  • Replacement parts for syringe with valve
  • Push-pull with needle
Specifications
For Use With (Equipment) 25 μL to 2.5 mL Removable Needle Syringes
Includes 5 cm L x 0.63 O.D. Replaceable Cone Tip Needle