Learn More
Abcam Purine Nucleoside Phosphorylase Activity Assay Kit (Fluorometric)
Product Code. 32120919 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
Product Code. Quantity Unit Size
32120919 100 Tests 100 tests
1 options
This item is not returnable. View return policy
Product Code. 32120919 Supplier Abcam Supplier No. ab204706100Test
 Restricted Item
Fecha estimada de envoi: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Measure Nucleoside phosphorylase activity in Cell/Tissue Extracts with Purine Nucleoside Phosphorylase Activity Assay Kit AB204706. Sensitivity = 0.005 μU. Cited in 1 publication. Use with Microplate reader.
Measure Nucleoside phosphorylase activity in Cell/Tissue Extracts with Purine Nucleoside Phosphorylase Activity Assay Kit AB204706. Sensitivity = 0.005 μU. Cited in 1 publication. Use with Microplate reader.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 100 Tests
Regulatory Status RUO