Description
Protects DRG1 from proteolytic degradation.
Specifications
Specifications
| Antigen | PTP1B |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.35 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PTPN1 |
| Gene Accession No. | P18031, P20417, P35821 |
| Gene Alias | PTP 1B, PTP1B, PTPN1 |
| Gene Symbols | PTPN1 |
| Show More |