Learn More
PTK2 (FAK) Recombinant Human Protein
PTK2 (FAK) Recombinant Human Protein
Marca:  Invitrogen PV3832
Detalles adicionales : Peso : 0.05250kg
 Ver más versiones de este producto
Código de producto. 11543567
  • 464.00€ / 10 microgramos
Fecha de envío estimada
del proveedor 07-02-2019
Añadir a la cesta
Descripción
Descripción
PTK2 (FAK) is a cytoplasmic protein tyrosine kinase which is found concentrated in the focal adhesions that form between cells growing in the presence of extracellular matrix constituents.
Especificación
Especificación
NP_722560
Cell viability, proliferation and function, pharma and biopharma, protein assays and analysis, target and lead identification and validation
146.7kDa
80% by SDS-PAGE
–80°C
1 Each
PTK2 (FAK), FADK, FAK1
PTK2 (FAK) was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.0mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 and trace [32P]-γ-ATP for 10 minutes at 30°C
Kinase and inhibitors
FAK
100nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 4μg/mL
GST Tag
0.46mg/mL
3725, 5747
PTK2 (FAK) Recombinant Human Protein
10μg
For research use only. Not for use in diagnostic procedures
Cytoplasmic tyrosine kinase
PTK2 (FAK) Recombinant Human Protein
Kinase assay
Baculovirus
PTK2 (FAK) Recombinant Human Protein
Human
Videos
For Research Use Only. Not for use in diagnostic procedures.