Learn More
PTH1R Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 29115-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
PTH1R is a receptor for parathyroid hormone and parathyroid hormone-related peptide (PTHrP). PTH1R is a glycoprotein with seven transmembrane domains and it has an extracellular N-terminus region and an intracellular C-terminus region. The activity of PTH1R is mediated by G proteins which activate adenylyl cyclase. It has been reported that PTHrP and PTH1R are associated with the differentiation of bone and cartilage in the developing stages. PTH1R has several isoforms with 66/59 kDa and PTH1R can form a 120 kDa dimer in the non-reducing condition.
Especificaciones
PTH1R | |
Polyclonal | |
Unconjugated | |
20 μL | |
5745 | |
Liquid |
Western Blot, Immunohistochemistry, ELISA | |
700 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse, Rat | |
IgG |