Learn More
Lenz Laborglasinstrumente™ Mezcladora de PTFE
Ideal for routine stirring and mixing applications. Lenz Laborglasinstrumente™ Stirrers with PTFE Blade and Journal feature a shaft manufactured from durable DURAN™ tubing.
Marca: Lenz Laborglasinstrumente™ B3972522
Detalles adicionales : Peso : 0.04500kg
Descripción
- Precision calibrated glass tube
- Ground and polished bearing surface
- With snap fastener for replacement of PTFE blades