Learn More
Fisherbrand™ PTFE Joint Adapter
Creates high vacuum with sealing rings, negating the need for grease. Cowie Technology™ PTFE Joint Adapter are autoclavable and reusable.
Marca:  Fisherbrand™
Detalles adicionales : Peso : 0.08000kg
 Ver más versiones de este producto
Código de producto. 11503342
  • 43.95€ / paquete de 2
Fecha de envío estimada
del proveedor 14-02-2019
Añadir a la cesta
Descripción
Descripción
Made from PTFE with a substantial shoulder for ease of handling and an overall robust construction for repeated use. Rigid sleeves and adapters are inert, usable to 280°C (approx.) and will withstand high vacuum. High vacuum is obtained using transverse trapezoidal sealing rings on the contacting surfaces. For grease free, high vacuum seals using standard ground glass joints.
Especificación
Especificación
1Each
19/26
PTFE Joint Adapters
Videos