Learn More
Fisherbrand™ PTFE Flat Head Stopper
Contains ‘B’ Length Cone for Europe. Fisherbrand™ PTFE Flat Head Stopper can be used up to 280°C.
Marca: Fisherbrand™
Detalles adicionales : Peso : 0.01400kg
Descripción
- Operates at high vacuum - transverse trapezoidal sealing rings
- Leakage Rate < 1 x 10-4 torr, litre, sec-1