Learn More
Fisherbrand™ Tapa de vaso de precipitados de PTFE
Click to view available options
Diameter: 100mm
Descripción
- Inert, chemically resistant
- Smooth internal finish
- Exceptional thermal resistance: can be used down to -200°C and up to 280°C
- Isostatically molded from pure PTFE