missing translation for 'onlineSavingsMsg'
Learn More

PSMD11 Antibody (1F4), HRP, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NBP1-30252H

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213762

  • 564.78€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

PSMD11 Monoclonal specifically detects PSMD11 in Human, Mouse, C. elegans samples. It is validated for Western Blot, ELISA.
Especificaciones

Especificaciones

PSMD11
Monoclonal
HRP
PBS with No Preservative
PSMD11
Recombinant human PSMD11 (1-422aa) purified from E. coli (NP_002806).
0.1 mL
Primary
This antibody is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human PSMD11 protein.
Store at 4C in the dark.
IgG1 κ
Western Blot, ELISA
1F4
Western Blot, ELISA
MGC3844,26S proteasome regulatory subunit RPN6, p44.5,26S proteasome regulatory subunit 9, proteasome (prosome, macropain) 26S subunit, non-ATPase, 11,26S proteasome regulatory subunit p44.5, Rpn6,26S proteasome regulatory subunit S9, S9,26S proteasome non-ATPase regulatory subunit 11
Mouse
Protein A or G purified
RUO
5717
Human, Mouse, C. elegans
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales