missing translation for 'onlineSavingsMsg'
Learn More

PSMD11 Antibody (1F4), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP1-30252AF750

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213593

  • 551.82€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

PSMD11 Monoclonal specifically detects PSMD11 in Human, Mouse, C. elegans samples. It is validated for Western Blot, ELISA.
Especificaciones

Especificaciones

PSMD11
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
PSMD11
Recombinant human PSMD11 (1-422aa) purified from E. coli (NP_002806).
0.1 mL
5717.0
Store at 4°C in the dark.
Western Blot, ELISA
1F4
Western Blot, ELISA
MGC3844,26S proteasome regulatory subunit RPN6, p44.5,26S proteasome regulatory subunit 9, proteasome (prosome, macropain) 26S subunit, non-ATPase, 11,26S proteasome regulatory subunit p44.5, Rpn6,26S proteasome regulatory subunit S9, S9,26S proteasome non-ATPase regulatory subunit 11
Mouse
Protein G purified
Primary
Human, Mouse, C. elegans
IgG1 κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales