missing translation for 'onlineSavingsMsg'
Learn More

PSMB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 15154-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16814214

  • 109.00€ / 20 microlitros

Envío estimado: 21-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the proteasome B-type family, also known as the T1B family, that is a 20S core beta subunit. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Especificaciones

Especificaciones

PSMB2
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P40307, P49721, Q9R1P3
20S proteasome, HC7 I, Macropain subunit C7 I, Proteasome component C7 I, Proteasome subunit beta type 2, PSMB2
Rabbit
IgG
20 μL
-20°C
Primary
26445, 29675, 5690
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
PSMB2
Liquid
PSMB2
PSMB2 Fusion Protein Ag7198
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Rat, Mouse, Human
Videos
SDS
Documentos
Certificados