missing translation for 'onlineSavingsMsg'
Learn More

PSAP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 18398-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16818464

  • 109.00€ / 20 microlitros

Envío estimado: 27-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This antibody should recognize saposin C.

PSAP is a highly conserved glycoprotein which is a precursor for 4 cleavage products: saposins A, B, C, and D. Each domain of the precursor protein is approximately 80 amino acid residues long with nearly identical placement of cysteine residues and glycosylation sites. Saposins A-D localize primarily to the lysosomal compartment where they facilitate the catabolism of glycosphingolipids with short oligosaccharide groups. The precursor protein exists both as a secretory protein and as an integral membrane protein and has neurotrophic activities.
Especificaciones

Especificaciones

PSAP
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P07602, P10960, Q61207
A1 activator, Cerebroside sulfate activator, Co beta glucosidase, Component C, CSAct, Dispersin, GLBA, Glucosylceramidase activator, Proactivator polypeptide, prosaposin, Protein C, PSAP, SAP 1, SAP 2 Saposin D, SAP1
Rabbit
IgG
20 μL
-20°C
Primary
19156, 25524, 5660
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
Unconjugated
PSAP
Liquid
Psap
PSAP Fusion Protein Ag13265
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Human, Rat
Videos
SDS
Documentos
Certificados