missing translation for 'onlineSavingsMsg'
Learn More

PRR7 Antibody (TRAP3/10), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP1-30424AF647

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18216260

  • 537.78€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

PRR7 Monoclonal specifically detects PRR7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Especificaciones

Especificaciones

PRR7
Monoclonal
Alexa Fluor 647
MGC10772, proline rich 7 (synaptic), proline-rich protein 7, Synaptic proline-rich membrane protein
Mouse
Protein A purified
RUO
80758
Human, Mouse, Rat
Purified
Western Blot, Immunocytochemistry, Immunofluorescence
TRAP3/10
Western Blot, Immunocytochemistry/Immunofluorescence
PRR7
Recombinant C-terminal half of the intracellular domain of human PRR7/TRAP3 (amino acids 126-253)
0.1 mL
Primary
The mouse monoclonal antibody TRAP3/10 recognizes an epitope located in the C-terminal part of the intracellular domain of PRR7/TRAP3 (amino acids 126-253 of human PRR7 TRAP3), a 28 kDa proline-rich membrane protein presumably associated with NMDA receptor complex.
Store at 4C in the dark.
IgG2a
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only