missing translation for 'onlineSavingsMsg'
Learn More

Terracycle™ Protective Eyewear Zero Waste Box™

Product Code. 17895695
Change view
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17895695 Medium Each
17815895 Small Each
17845895 Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17895695 Supplier Terracycle™ Supplier No. CS93MES
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recycle Protective Eyewear using this Zero Waste Box system. Zero Waste Box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.

Use this Zero Waste Box to recycle:

  • Swimming goggles, tanning and safety glasses

Approximate waste units accepted:

  • Small: 200 units
  • Medium: 510 units
  • Large: 1000 units

Do not send in:

  • Sunscreen bottles
  • Other protective equipment
  • Medical waste
    • This includes material contaminated with blood or bodily fluids and waste that originate from health care facilities, hospitals, home care, physicians' offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.
  • Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) which presents a danger to the environment, or to people
  • Batteries, pressurized canisters, broken glass and medical waste

Specifications

Capacity (Metric) 15 kg
Capacity (English) 33 lb.
Color Beige, Green, Red, White
Dimensions (L x W x H) 30.5 x 30.5 x 84 cm
Disposable Single-use
Latex Free Yes
Size Medium
Sterility Non-sterile
Autoclavable Not Autoclavable
Material Cardboard, Plastic
Product Type Recycling Box
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.