missing translation for 'onlineSavingsMsg'
Learn More

Progesterone Receptor Ab-7, Mouse Monoclonal Antibody,Lab Vision™ Epredia™

Provide accurate, reproducible results with the Epredia™ Progesterone Receptor Ab-7, Mouse Monoclonal Antibody.

Marca:  Epredia MS-197-P0

Detalles adicionales : Peso : 1.00000kg

 Ver más versiones de este producto

Código de producto. 12670497

  • 136.00€ / 100 microlitros

Envío estimado: 29-06-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Human PgR exists in two forms: 116kDa (B-form) and 81kDa (A-form). It acts as ligand activated transcription factor to regulate expression of the target genes. Null mutation in PgR gene leads to pleiotrophic reproductive abnormalities.

Host Species: Mouse

Clone: hPRa 7

Isotype: IgG1/κ

Species Reactivity: Human, Baboon, Monkey, Mouse, and Rat. Does not react with guinea pig. Others not known.

Epitope: N-terminal half of human PgR

Immunogen: PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice

Molecular Weight: 116kDa (triplet, B-form) and 81kDa (singlet, A-form)

Positive Control: T47D cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native only)
  • Sedimentation in Sucrose Gradient

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

Progesterone Receptor Ab-7
Monoclonal
200μg/mL
Concentrated
PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice
100μL
Cancer and Tumor Biology
Primary
Human, Murine, Non-human Primate, Rat
Immunofluorescence, Immunoprecipitation, Western Blot
hPRa 7
Unconjugated
Murine
IgG2a kappa
RUO
Primary
Monoclonal
Videos
SDS
Documentos
Certificados